Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human CTSG native Protein from human neutrophils

Used for Func, SDS-PAGE

Manufacturer:  Abnova™ P4914

Catalog No. 89-940-941


Add to cart

Description

Description

The protein encoded by this gene, a member of the peptidase S1 protein family, is found in azurophil granules of neutrophilic polymorphonuclear leukocytes. The encoded protease has a specificity similar to that of chymotrypsin C, and may participate in the killing and digestion of engulfed pathogens, and in connective tissue remodeling at sites of inflammation. Transcript variants utilizing alternative polyadenylation signals exist for this gene. [provided by RefSeq]

Specifications

Specifications

0.2 mg/mL
Liquid
1511
CTSG (Human) Native Protein
Saltprecipitation/Affinity Chromatography/Ion Exchange Chromatography/Gelfiltration
15% SDS-PAGE Stained with Coomassie Blue
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
CG|MGC23078
CTSG
∽3000mU/mg
None
Functional Study, Sodium Dodecyl Sulfate–Polyacrylamide Gel Electrophoresis
In 50mM acetate, 250mM NaCl, pH 5.5
29kDa
Native protein purified from human neutrophils
>95% by SDS-PAGE
25 ug
RUO
CTSG
Native protein purified from human neutrophils
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit