Learn More
Abnova™ Human CTSH (P09668) native Protein from human liver
Used for Func, SDS-PAGE
Manufacturer:  Abnova™ P4031
Catalog No. 89-940-948
Add to cart
Description
Description
The protein encoded by this gene is a lysosomal cysteine proteinase important in the overall degradation of lysosomal proteins. It is composed of a dimer of disulfide-linked heavy and light chains, both produced from a single protein precursor. The encoded protein, which belongs to the peptidase C1 protein family, can act both as an aminopeptidase and as an endopeptidase. Increased expression of this gene has been correlated with malignant progression of prostate tumors. Two transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq]