Learn More
Abnova™ Human CUGBP2 Full-length ORF (AAH36391.1, 1 a.a. - 521 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Supplier: Abnova™ H00010659P01S
Description
Members of the CELF/BRUNOL protein family contain two N-terminal RNA recognition motif (RRM) domains, one C-terminal RRM domain, and a divergent segment of 160-230 aa between the second and third RRM domains. Members of this protein family regulate pre-mRNA alternative splicing and may also be involved in mRNA editing, and translation. Alternative splicing results in multiple transcript variants encoding different isoforms. [provided by RefSeq]
Sequence: MMVEGRLLVPDRINGTANKMNGALDHSDQPDPDAIKMFVGQIPRSWSEKELKELFEPYGAVYQINVLRDRSQNPPQSKGCCFVTFYTRKAALEAQNALHNIKTLPGMHHPIQMKPADSEKSNAVEDRKLFIGMVSKKCNENDIRVMFSPFGQIEECRILRGPDGLSRGCAFVTFSTRAMAQNAIKAMHQSQTMEGCSSPIVVKFADTQKDKEQRRLQQQLAQQMQQLNTATWGNLTGLGGLTPQYLALLQQATSSSNLGAFSGIQQMAGMNALQLQNLATLAAAAAAAQTSATSTNANPLSTTSSALGALTSPVAASTPNSTAGAAMNSLTSLGTLQGLAGATVGLNNINALAGTINTPRSKRLLLPKDNNGMAALNGGLGATGLTNGTAGTMDALTQAYSGIQQYAAAALPTLYSQSLLQQQSAAGSQKEGPEGANLFIYHLPQEFGDQDILQMFMPFGNVISAKVFIDKQTNLSKCFGFVSYDNPVSAQAAIQAMNGFQIGMKRLKVQLKRSKNDSKPYSpecifications
AAH36391.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
82.2kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
BRUNOL3/ETR-3/NAPOR | |
CUGBP2 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
10659 | |
CUGBP2 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MMVEGRLLVPDRINGTANKMNGALDHSDQPDPDAIKMFVGQIPRSWSEKELKELFEPYGAVYQINVLRDRSQNPPQSKGCCFVTFYTRKAALEAQNALHNIKTLPGMHHPIQMKPADSEKSNAVEDRKLFIGMVSKKCNENDIRVMFSPFGQIEECRILRGPDGLSRGCAFVTFSTRAMAQNAIKAMHQSQTMEGCSSPIVVKFADTQKDKEQRRLQQQLAQQMQQLNTATWGNLTGLGGLTPQYLALLQQATSSSNLGAFSGIQQMAGMNALQLQNLATLAAAAAAAQTSATSTNANPLSTTSSALGALTSPVAASTPNSTAGAAMNSLTSLGTLQGLAGATVGLNNINALAGTINTPRSKRLLLPKDNNGMAALNGGLGATGLTNGTAGTMDALTQAYSGIQQYAAAALPTLYSQSLLQQQSAAGSQKEGPEGANLFIYHLPQEFGDQDILQMFMPFGNVISAKVFIDKQTNLSKCFGFVSYDNPVSAQAAIQAMNGFQIGMKRLKVQLKRSKNDSKPY | |
RUO | |
CUGBP2 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.