Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human CX36 Partial ORF (NP_065711.1, 99 a.a. - 197 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Manufacturer:  Abnova™ H00057369Q01S

 View more versions of this product

Catalog No. 89-941-014


Add to cart

Description

Description

This gene is a member of the connexin gene family that is expressed predominantly in mammalian neurons. Connexins associate in groups of 6 and are organized radially around a central pore to form connexons. Each gap junction intercellular channel is formed by the conjunction of 2 connexons. [provided by RefSeq]

Sequence: HQSAKQRERRYSTVFLALDRDPPESIGGPGGTGGGGSGGGKREDKKLQNAIVNGVLQNTENTSKETEPDCLEVKELTPHPSGLRTASKSKLRRQEGISR
Specifications

Specifications

NP_065711.1
Liquid
57369
CX36 (Human) Recombinant Protein (Q01)
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
CX36|GJA9|MGC138315|MGC138319
GJD2
Wheat Germ (in vitro)
GST
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.63kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
HQSAKQRERRYSTVFLALDRDPPESIGGPGGTGGGGSGGGKREDKKLQNAIVNGVLQNTENTSKETEPDCLEVKELTPHPSGLRTASKSKLRRQEGISR
RUO
GJD2
wheat germ expression system
Yes
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit