Learn More
Abnova™ Human CXCL10 (P02778, 22 a.a. - 98 a.a.) Partial Recombinant Protein
Used for Func, SDS-PAGE
Manufacturer:  Abnova™ P3643
Catalog No. 89-941-037
Add to cart
Description
Description
This gene encodes a chemokine of the CXC subfamily and ligand for the receptor CXCR3. Binding of this protein to CXCR3 results in pleiotropic effects, including stimulation of monocytes, natural killer and T-cell migration, and modulation of adhesion molecule expression. [provided by RefSeq]
Sequence: VPLSRTVRCTCISISNQPVNPRSLEKLEIIPASQFCPRVEIIATMKKKGEKRCLNPESKAIKNLLKAVSKERSKRSP