Learn More
Abnova™ Human CXCL12 (beta) (P48061-1) Recombinant Protein
Used for Func, SDS-PAGE
Supplier: Abnova™ P4566
Description
For background information on chemokines, see CXCL1 (MIM 155730). Stromal cell-derived factors 1-alpha and 1-beta are small cytokines that belong to the intercrine family, members of which activate leukocytes and are often induced by proinflammatory stimuli such as lipopolysaccharide, TNF (see MIM 191160), or IL1 (see MIM 147760). The intercrines are characterized by the presence of 4 conserved cysteines which form 2 disulfide bonds. They can be classified into 2 subfamilies. In the CC subfamily, which includes beta chemokine, the cysteine residues are adjacent to each other. In the CXC subfamily, which includes alpha chemokine, they are separated by an intervening amino acid. The SDF1 proteins belong to the latter group.[supplied by OMIM]
Sequence: KPVSLSYRCPCRFFESHVARANVKHLKILNTPNCALQIVARLKNNNRQVCIDPKLKWIQEYLEKALNKRFKMSpecifications
P48061-1 | |
Lyophilized | |
8.5kDa | |
Escherichia coli expression system | |
10 ug | |
Store at -20°C on dry atmosphere. After reconstitution with sterilized water, store at -20°C. Aliquot to avoid repeated freezing and thawing. | |
<0.1 EU/μg | |
CXCL12 | |
The activity is determined by the ability to chemoattract human peripheral T cells and is detectable starting at 5ng/mL. | |
Recombinant | |
Escherichia coli expression system |
Functional Study, SDS-PAGE | |
6387 | |
CXCL12 (Beta) (Human) Recombinant Protein | |
1 ug/lane in 4-20% Tris-Glycine gel Stained with Coomassie Blue | |
KPVSLSYRCPCRFFESHVARANVKHLKILNTPNCALQIVARLKNNNRQVCIDPKLKWIQEYLEKALNKRFKM | |
RUO | |
PBSF/SCYB12/SDF-1a/SDF-1b/SDF1/SDF1A/SDF1B/TLSF-a/TLSF-b/TPAR1 | |
CXCL12 | |
E. coli | |
None | |
Lyophilized |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.