Learn More
Abnova™ Human CXCL13 (Q53X90, Val 22 - Phe 109) Partial Recombinant Protein
Used for Func, SDS-PAGE
Supplier: Abnova™ P3589
Description
B lymphocyte chemoattractant, independently cloned and named Angie, is a CXC chemokine strongly expressed in the follicles of the spleen, lymph nodes, and Peyer's patches. It preferentially promotes the migration of B lymphocytes (compared to T cells and macrophages), apparently by stimulating calcium influx into, and chemotaxis of, cells expressing Burkitt's lymphoma receptor 1 (BLR-1). It may therefore function in the homing of B lymphocytes to follicles. [provided by RefSeq]
Sequence: Val 22 - Phe 109Specifications
Q53X90 | |
Lyophilized | |
10kDa | |
Escherichia coli expression system | |
20 ug | |
Store at -20°C on dry atmosphere for 2 years. After reconstitution with deionized water, store at 4°C for 1 month or store at -20°C for 6 months. Aliquot to avoid repeated freezing and thawing. | |
<0.1ng/μg (1 EU/μg) | |
CXCL13 | |
Determined by its ability to chemoattract human T cells using a concentration range of 2.0-10.0ng/mL. | |
Recombinant | |
Escherichia coli expression system | |
>90% by SDS-PAGE and HPLC |
Functional Study, SDS-PAGE | |
10563 | |
CXCL13 (Human) Recombinant Protein | |
Ion exchange column and HPLC reverse phase column | |
Val 22 - Phe 109 | |
RUO | |
ANGIE/ANGIE2/BCA-1/BCA1/BLC/BLR1L/SCYB13 | |
CXCL13 | |
E. coli | |
None | |
Lyophilized |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.