Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human CXCL5/ENA-78 Alexa Fluor 488-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems IC654G100UG
This item is not returnable.
View return policy
Description
CXCL5/ENA-78 Monoclonal specifically detects CXCL5/ENA-78 in Human samples. It is validated for Intracellular Staining by Flow Cytometry.Specifications
CXCL5/ENA-78 | |
Monoclonal | |
0.2 mg/mL | |
Intracellular Staining by Flow Cytometry 0.25-1 ug/10^6 cells | |
P42830 | |
CXCL5 | |
E. coli-derived recombinant human CXCL5/ENA-78 Ala37-Asn114 Accession # P42830 | |
RUO | |
6374 | |
Human | |
IgG1 |
Flow Cytometry | |
33170 | |
Alexa Fluor 488 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
chemokine (C-C motif) ligand 5, C-X-C motif chemokine 5, CXCL5, ENA78, ENA-78, ENA-78(1-78), Epithelial-derived neutrophil-activating protein 78, Neutrophil-activating peptide ENA-78, neutrophil-activating protein 78, SCYB5ENA78 | |
Mouse | |
100 μg | |
Primary | |
Detects human CXCL5/ENA-78 in ELISAs. Does not cross-react with recombinant human CXCL1, 2, or 3. | |
Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction