Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Abnova™ Human CXCL5 (P42830, 74 amino acids) Partial Recombinant Protein
Used for Func, SDS-PAGE
Supplier: Abnova™ P3961
Description
The protein encoded by this gene is an inflammatory chemokine that belongs to the CXC chemokine family. This chemokine is produced concomitantly with interleukin-8 (IL8) in response to stimulation with either interleukin-1 (IL1) or tumor necrosis factor-alpha (TNFA). This chemokine is a potent chemotaxin involved in neutrophil activation. [provided by RefSeq]
Specifications
P42830 | |
Lyophilized | |
8kDa | |
Escherichia coli expression system | |
Store at -20°C on dry atmosphere. After reconstitution with deionized water, store at -20°C. Aliquot to avoid repeated freezing and thawing. | |
<0.1ng/μg | |
CXCL5 | |
The human ENA-78 produces chemotaxis over a wide concentration range in an assay using human peripheral blood neutrophils. Significant chemotaxis was achieved using a concentration of 50ng/mL. | |
Recombinant | |
Escherichia coli expression system | |
≥98% by SDS-PAGE and HPLC |
Functional Study, SDS-PAGE | |
6374 | |
CXCL5 (Human) Recombinant Protein | |
20 ug | |
RUO | |
ENA-78/SCYB5 | |
CXCL5 | |
E. coli | |
None | |
Lyophilized |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction