Learn More
Abnova™ Human CXCL5 (P42830) Partial Recombinant Protein
Used for Func, SDS-PAGE
Manufacturer:  Abnova™ P5972
Catalog No. 89-941-077
Add to cart
Description
Description
The protein encoded by this gene is an inflammatory chemokine that belongs to the CXC chemokine family. This chemokine is produced concomitantly with interleukin-8 (IL8) in response to stimulation with either interleukin-1 (IL1) or tumor necrosis factor-alpha (TNFA). This chemokine is a potent chemotaxin involved in neutrophil activation. [provided by RefSeq]
Sequence: AAVLRELRCVCLQTTQGVHPKMISNLQVFAIGPQCSKVEVVASLKNGKEICLDPEAPFLKKVIQKILDGGNKEN