Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human CXCL9 (aa 23-125), Recombinant Protein, Invitrogen™ Non-distribution product as customer accommodation.

E. coli Recombinant Protein

$160.50 - $3090.00

Specifications

Recombinant Yes
Name Human CXCL9 (aa 23-125)
For Use With (Application) Control, ELISA, Western Blot
Concentration 200 µg/mL
Conjugate Unlabeled
View More Specs

Products 4
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
PIRP87841 Non-distribution product as customer accommodation.
View Documents
Thermo Scientific
RP87841
10 µg
Each for $160.50
Add to cart
 
PIRP87842 Non-distribution product as customer accommodation.
View Documents
Thermo Scientific
RP87842
25 µg
Each for $289.00
Add to cart
 
PIRP87843 Non-distribution product as customer accommodation.
View Documents
Thermo Scientific
RP87843
100 µg
Each for $764.00
Add to cart
 
PIRP87844 Non-distribution product as customer accommodation.
View Documents
Thermo Scientific
RP87844
500 µg
Each for $3,090.00
Add to cart
 
Description

Description

MIG, also known as CXCL9, is a T-cell chemoattractant inducible by gamma interferon that is a member of the CXC chemokine family of cytokines.This gene is mapped to 4q21. It is noted that, although the best-described activities of the chemokines are as chemotactic factors, chemokines also have an effect on T-cell activation, angiogenesis, and HIV infection. While most CXC chemokines are chemotactic for neutrophils, MIG and INP10 are unusual and similar in being CXC chemokines that are chemotactic for lymphocytes and inactive in neutrophils.
Specifications

Specifications

Yes
Control, ELISA, Western Blot
Unlabeled
PBS with no preservative
E. coli
-20°C
Q07325
CXCL9
>95%
Human CXCL9 (aa 23-125)
200 µg/mL
Liquid
BB139920; C Cmotif chemokine; C x C motif chemokine; CC motif chemokine; CCmotif chemokine; chemokine (C-X-C motif) ligand 9; CMK; crg-10; CXC; CXC motif chemokine; C-X-C motif chemokine 9; C-X-C motif chemokine ligand 9; CXCL; Cxcl9; gamma interferon-induced monokine; gamma-interferon-induced monokine; HuMIG; M119; MIG; monokine induced by gamma interferon; monokine induced by interferon gamma; monokine induced by interferon-gamma; MuMIG; Protein m119; SCYB9; small inducible cytokine B subfamily (Cys-X-Cys), member 9; small inducible cytokine B9; small-inducible cytokine B9
RUO
Human
4283
11.7 kD
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit