Learn More
Abnova™ Human CXCL9 (Q07325, 23 a.a. - 125 a.a.) Partial Recombinant Protein
Used for Func, SDS-PAGE
Manufacturer:  Abnova™ P3650
Catalog No. 89-941-089
Add to cart
Description
Description
The function of this gene has not been specifically defined; however, it is thought to be involved in T cell trafficking. This gene has been localized to 4q21 with INP10, which is also a member of the chemokine family of cytokines. [provided by RefSeq]
Sequence: TPVVRKGRCSCISTNQGTIHLQSLKDLKQFAPSPSCEKIEIIATLKNGVQTCLNPDSADVKELIKKWEKQVSQKKKQKNGKKHQKKKVLKVRKSQRSRQKKTT