Learn More
Novus Biologicals™ Human CXCR1/IL-8RA ELISA Kit (Colorimetric)
Colorimetric ELISA Kit
Supplier: Novus Biologicals™ NBP306836
This item is not returnable.
View return policy
Specifications
0.31 to 20 ng/mL | |
ELISA Kit (Colorimetric) | |
This kit recognizes Human CXCR1/IL-8RA in samples. No significant cross-reactivity or interference between Human CXCR1/IL-8RA and analogues was observed. | |
3577 | |
1 Kit | |
Storage of components varies. See protocol for specific instructions. | |
Human |
0.19 ng/mL | |
Biological Fluids, Cell Culture Supernatant, Plasma, Serum | |
ELISA, Sandwich ELISA | |
CXCR1 | |
Chemokines and Cytokines, Cytokine Research, GPCR, Immunology, Innate Immunity, Signal Transduction | |
CD128, CD181, CD181 antigen, CDw128aC-C, chemokine (C-X-C motif) receptor 1, CKR-1, CMKAR1, C-X-C chemokine receptor type 1, CXC-R1, CXCR-1, High affinity interleukin-8 receptor A, IL-8 receptor type 1, IL-8R A, IL8R1, IL8RAC-C-CKR-1, IL8RBA, interleukin 8 receptor, alpha, interleukin-8 receptor type 1, interleukin-8 receptor type A |