Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human CXCR3 Partial ORF (NP_001495.1, 1 a.a. - 53 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Manufacturer:  Abnova™ H00002833Q02L

 View more versions of this product

Catalog No. 89-941-100


Add to cart

Description

Description

This gene encodes a G pr-coupled receptor with selectivity for 3 chemokines, termed IP10, Mig and I-TAC. IP10, Mig and I-TAC belong to the structural subfamily of CXC chemokines, in which a single aa residue separates first 2 of 4 highly conserved Cys residues. Binding of chemokines to this pr induces cellular responses that are involved in leukocyte traffic, most notably integrin activation, cytoskeletal changes and chemotactic migration. Inhibition by Bordetella pertussis toxin suggests that heterotrimeric G pr of the Gi-subclass couple to this protein. Signal transduction may include same enzymes that were identified in the signaling cascade induced by other chemokine receptors. As a consequence of chemokine-induced cellular desensitization, cellular responses are typically rapid and short in duration. Cellular responsiveness is restored after dephosphorylation of intracellular receptors and subsequent recycling to cell surface. This gene is prominently expressed in in vitro cultured effector/memory T cells, and in T cells present in many types of inflamed tissues. In addition, IP10, Mig and I-TAC are commonly produced by local cells in inflammatory lesion, suggesting that this gene and its chemokines participate in recruitment of inflammatory cells. So, this protein is a target for development of small M.W. antagonists, which may be used in treatment of diverse inflammatory diseases. Multiple transcript variants encoding different isoforms have been found for this gene.

Sequence: MVLEVSDHQVLNDAEVAALLENFSSSYDYGENESDSCCTSPPCPQDFSLNFDR
Specifications

Specifications

NP_001495.1
Liquid
2833
CXCR3 (Human) Recombinant Protein (Q02)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
CD182|CD183|CKR-L2|CMKAR3|GPR9|IP10-R|Mig-R|MigR
CXCR3
Wheat Germ (in vitro)
GST
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
31.57kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
MVLEVSDHQVLNDAEVAALLENFSSSYDYGENESDSCCTSPPCPQDFSLNFDR
RUO
CXCR3
wheat germ expression system
Yes
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit