Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human CYB5R4 Partial ORF (NP_057314, 388 a.a. - 487 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Manufacturer:  Abnova™ H00051167Q01S

 View more versions of this product

Catalog No. 89-953-683


Add to cart

Description

Description

NCB5OR is a flavohemoprotein that contains functional domains found in both cytochrome b5 (CYB5A; MIM 250790) and CYB5 reductase (see MIM 250800).[supplied by OMIM]

Sequence: VKLMFFNKTEDDIIWRSQLEKLAFKDKRLDVEFVLSAPISEWNGKQGHISPALLSEFLKRNLDKSKVLVCICGPVPFTEQGVRLLHDLNFSKNEIHSFTA
Specifications

Specifications

wheat germ expression system
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.74kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
VKLMFFNKTEDDIIWRSQLEKLAFKDKRLDVEFVLSAPISEWNGKQGHISPALLSEFLKRNLDKSKVLVCICGPVPFTEQGVRLLHDLNFSKNEIHSFTA
RUO
CYB5R4
Wheat Germ (in vitro)
GST
NP_057314
Liquid
51167
CYB5R4 (Human) Recombinant Protein (Q01)
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
NCB5OR/RP4-676J13.1/dJ676J13.1
CYB5R4
Yes
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit