Learn More
Novus Biologicals™ Human Cyclin D2 ELISA Kit (Colorimetric)
ELISA kits are ideal for the quantification of target antigens in tissue extracts, serum and cell culture.
Manufacturer:  Novus Biologicals™ NBP275103
Catalog No. NBP275103
Add to cart
Description
Description
The Human Cyclin D2 ELISA Kit (Colorimetric) measures human Cyclin D2 in serum, plasma and other biological fluids.
Specifications
Specifications
0.16 - 10 ng/mL
ELISA
ELISA Kit (Colorimetric)
Storage is content dependent.
Human
1 Kit
This kit recognizes Human CCND2 in samples. No significant cross-reactivity or interference between Human CCND2 and analogues was observed.
894
0.10 ng/mL
Cyclin D2
CCND2
Biological Fluids, Plasma, Serum