Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human CYP2C18 Partial ORF (NP_000763, 200 a.a. - 299 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Manufacturer:  Abnova™ H00001562Q01L

 View more versions of this product

Catalog No. 89-941-235


Add to cart

Description

Description

This gene encodes a member of the cytochrome P450 superfamily of enzymes. The cytochrome P450 proteins are monooxygenases which catalyze many reactions involved in drug metabolism and synthesis of cholesterol, steroids and other lipids. This protein localizes to the endoplasmic reticulum but its specific substrate has not yet been determined. The gene is located within a cluster of cytochrome P450 genes on chromosome 10q24. An additional gene, CYP2C17, was once thought to exist; however, CYP2C17 is now considered an artefact based on a chimera of CYP2C18 and CYP2C19. Alternatively spliced transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq]

Sequence: KFNENLRILSSPWIQVCNNFPALIDYLPGSHNKIAENFAYIKSYVLERIKEHQESLDMNSARDFIDCFLIKMEQEKHNQQSEFTVESLIATVTDMFGAGT
Specifications

Specifications

NP_000763
Liquid
1562
CYP2C18 (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
CPCI|CYP2C|CYP2C17|DKFZp686I24235|P450-6B/29C|P450IIC17
CYP2C18
Wheat Germ (in vitro)
GST
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.74kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
KFNENLRILSSPWIQVCNNFPALIDYLPGSHNKIAENFAYIKSYVLERIKEHQESLDMNSARDFIDCFLIKMEQEKHNQQSEFTVESLIATVTDMFGAGT
RUO
CYP2C18
wheat germ expression system
Yes
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit