Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human CYP39A1 Full-length ORF (AAH10358, 1 a.a. - 469 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Manufacturer:  Abnova™ H00051302P01

Catalog No. 89-941-258


Add to cart

Description

Description

This gene encodes a member of the cytochrome P450 superfamily of enzymes. The cytochrome P450 proteins are monooxygenases which catalyze many reactions involved in drug metabolism and synthesis of cholesterol, steroids and other lipids. This endoplasmic reticulum protein is involved in the conversion of cholesterol to bile acids. Its substrates include the oxysterols 25-hydroxycholesterol, 27-hydroxycholesterol and 24-hydroxycholesterol. [provided by RefSeq]

Sequence: MELISPTVIIILGCLALFLLLQPKNLRRPPCIKGWIPWIGVGFEFGKAPLEFIEKARIKYGPIFTVFAMGNRMTFVTEEEGINVFLKSKKVDFELAVQNIVYRTASIPKNVFLALHEKLYIMLKGKMGTVNLHQFTGQLTEELHEQLENLGTHGTMDLNNLVRHLLYPVTVNMLFNKSLFSTNKKKIKEFHQYFQVYDEDFEYGSQLPECLLRNWSKSKKWFLELFEKNIPDIKACKSAKDNSMTLLQATLDIVETETSKENSPNYGLLLLWASLSNAVPVAFWTLAHVLSHPDIHKAIMEGISSVFGKAGKDKIKVSEDDLENLLLIKWCVLETIRLKAPGVITRKVVKPVEILNYIIPSGDLLMLSPFWLHRNPKYFPEPELFKPERWKKANLEKHSFLDCFMAFGSGKFQCPARWFALLEVQMCIILILYKYDCSLLDPLPKQSYLHLVGVPQPEGQCRIEYKQRI
Specifications

Specifications

AAH10358
Liquid
51302
CYP39A1 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MELISPTVIIILGCLALFLLLQPKNLRRPPCIKGWIPWIGVGFEFGKAPLEFIEKARIKYGPIFTVFAMGNRMTFVTEEEGINVFLKSKKVDFELAVQNIVYRTASIPKNVFLALHEKLYIMLKGKMGTVNLHQFTGQLTEELHEQLENLGTHGTMDLNNLVRHLLYPVTVNMLFNKSLFSTNKKKIKEFHQYFQVYDEDFEYGSQLPECLLRNWSKSKKWFLELFEKNIPDIKACKSAKDNSMTLLQATLDIVETETSKENSPNYGLLLLWASLSNAVPVAFWTLAHVLSHPDIHKAIMEGISSVFGKAGKDKIKVSEDDLENLLLIKWCVLETIRLKAPGVITRKVVKPVEILNYIIPSGDLLMLSPFWLHRNPKYFPEPELFKPERWKKANLEKHSFLDCFMAFGSGKFQCPARWFALLEVQMCIILILYKYDCSLLDPLPKQSYLHLVGVPQPEGQCRIEYKQRI
RUO
CYP39A1
Wheat Germ (in vitro)
GST
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
77.33kDa
Glutathione Sepharose 4 Fast Flow
2 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
CYP39A1
wheat germ expression system
Yes
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit