Learn More
Abnova™ Human CYP4F3 Partial ORF (NP_000887, 100 a.a. - 198 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Supplier: Abnova™ H00004051Q01S
Description
This gene, CYP4F3, encodes a member of the cytochrome P450 superfamily of enzymes. The cytochrome P450 proteins are monooxygenases which catalyze many reactions involved in drug metabolism and synthesis of cholesterol, steroids and other lipids. This protein localizes to the endoplasmic reticulum. The enzyme starts the process of inactivating and degrading leukotriene B4, a potent mediator of inflammation. This gene is part of a cluster of cytochrome P450 genes on chromosome 19. Another member of this family, CYP4F8, is approximately 18 kb away. [provided by RefSeq]
Sequence: RIFHPTYIKPVLFAPAAIVPKDKVFYSFLKPWLGDGLLLSAGEKWSRHRRMLTPAFHFNILKPYMKIFNESVNIMHAKWQLLASEGSARLDMFEHISLMSpecifications
NP_000887 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.63kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
RIFHPTYIKPVLFAPAAIVPKDKVFYSFLKPWLGDGLLLSAGEKWSRHRRMLTPAFHFNILKPYMKIFNESVNIMHAKWQLLASEGSARLDMFEHISLM | |
RUO | |
CYP4F3 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
4051 | |
CYP4F3 (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
CPF3/CYP4F/LTB4H | |
CYP4F3 | |
Recombinant | |
wheat germ expression system |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.