Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human CYP7B1 Full-length ORF (NP_004811.1, 1 a.a. - 506 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Manufacturer:  Abnova™ H00009420P01L

 View more versions of this product

Catalog No. 89-941-293


Add to cart

Description

Description

This gene encodes a member of the cytochrome P450 superfamily of enzymes. The cytochrome P450 proteins are monooxygenases which catalyze many reactions involved in drug metabolism and synthesis of cholesterol, steroids and other lipids. This endoplasmic reticulum membrane protein catalyzes the first reaction in the cholesterol catabolic pathway of extrahepatic tissues, which converts cholesterol to bile acids. This enzyme likely plays a minor role in total bile acid synthesis, but may also be involved in the development of atherosclerosis, neurosteroid metabolism and sex hormone synthesis. [provided by RefSeq]

Sequence: MAGEVSAATGRFSLERLGLPGLALAAALLLLALCLLVRRTRRPGEPPLIKGWLPYLGVVLNLRKDPLRFMKTLQKQHGDTFTVLLGGKYITFILDPFQYQLVIKNHKQLSFRVFSNKLLEKAFSISQLQKNHDMNDELHLCYQFLQGKSLDILLESMMQNLKQVFEPQLLKTTSWDTAELYPFCSSIIFEITFTTIYGKVIVCDNNKFISELRDDFLKFDDKFAYLVSNIPIELLGNVKSIREKIIKCFSSEKLAKMQGWSEVFQSRQDVLEKYYVHEDLEIGAHHLGFLWASVANTIPTMFWAMYYLLRHPEAMAAVRDEIDRLLQSTGQKKGSGFPIHLTREQLDSLICLESSIFEALRLSSYSTTIRFVEEDLTLSSETGDYCVRKGDLVAIFPPVLHGDPEIFEAPEEFRYDRFIEDGKKKTTFFKRGKKLKCYLMPFGTGTSKCPGRFFALMEIKQLLVILLTYFDLEIIDDKPIGLNYSRLLFGIQYPDSDVLFRYKVKS
Specifications

Specifications

NP_004811.1
Liquid
9420
CYP7B1 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MAGEVSAATGRFSLERLGLPGLALAAALLLLALCLLVRRTRRPGEPPLIKGWLPYLGVVLNLRKDPLRFMKTLQKQHGDTFTVLLGGKYITFILDPFQYQLVIKNHKQLSFRVFSNKLLEKAFSISQLQKNHDMNDELHLCYQFLQGKSLDILLESMMQNLKQVFEPQLLKTTSWDTAELYPFCSSIIFEITFTTIYGKVIVCDNNKFISELRDDFLKFDDKFAYLVSNIPIELLGNVKSIREKIIKCFSSEKLAKMQGWSEVFQSRQDVLEKYYVHEDLEIGAHHLGFLWASVANTIPTMFWAMYYLLRHPEAMAAVRDEIDRLLQSTGQKKGSGFPIHLTREQLDSLICLESSIFEALRLSSYSTTIRFVEEDLTLSSETGDYCVRKGDLVAIFPPVLHGDPEIFEAPEEFRYDRFIEDGKKKTTFFKRGKKLKCYLMPFGTGTSKCPGRFFALMEIKQLLVILLTYFDLEIIDDKPIGLNYSRLLFGIQYPDSDVLFRYKVKS
RUO
CYP7B1
wheat germ expression system
Yes
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
84.7kDa
Glutathione Sepharose 4 Fast Flow
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
CBAS3|CP7B|SPG5A
CYP7B1
Wheat Germ (in vitro)
GST
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit