Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human CYTL1 Partial ORF (NP_061129, 37 a.a. - 136 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Manufacturer:  Abnova™ H00054360Q01S

 View more versions of this product

Catalog No. 89-941-308


Add to cart

Description

Description

C17 is a cytokine-like protein specifically expressed in bone marrow and cord blood mononuclear cells that bear the CD34 (MIM 142230) surface marker (Liu et al., 2000 [PubMed 10857752]).[supplied by OMIM]

Sequence: EITRDFNLLQVSEPSEPCVRYLPRLYLDIHNYCVLDKLRDFVASPPCWKVAQVDSLKDKARKLYTIMNSFCRRDLVFLLDDCNALEYPIPVTTVLPDRQR
Specifications

Specifications

NP_061129
Liquid
54360
CYTL1 (Human) Recombinant Protein (Q01)
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
C17|C4orf4
CYTL1
Wheat Germ (in vitro)
GST
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.74kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
EITRDFNLLQVSEPSEPCVRYLPRLYLDIHNYCVLDKLRDFVASPPCWKVAQVDSLKDKARKLYTIMNSFCRRDLVFLLDDCNALEYPIPVTTVLPDRQR
RUO
CYTL1
wheat germ expression system
Yes
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit