Learn More
Novus Biologicals™ Human Cytochrome c ELISA Kit
ELISA kits are ideal for the quantification of target antigens in tissue extracts, serum and cell culture. 
Manufacturer:  Novus Biologicals™ NBP191263
Catalog No. NBP191263
Add to cart
Description
Description
The Human Cytochrome c ELISA Kit (Colorimetric) measures human Cytochrome c in cell culture supernatant, serum, plasma (edta, heparin), amniotic fluid.
Specifications
Specifications
0.08 - 5.0ng/mL
54205
ELISA Kit
0.05ng/mL
Human (Hu)
Cytochrome c
CYCS
ELISA
Aluminium pouch(es) with a Microwell Plate coated with monoclonal antibody to human Cytochrome c, Biotin-Conjugate anti-human Cytochrome c monoclonal antibody, Streptavidin-HRP, Human Cytochrome c Standard lyophilized, 10ng/mL upon reconstitution, Assay Buffer Concentrate 20x (PBS with 1% Tween 20 and 10% BSA), Wash Buffer Concentrate 20x (PBS with 1% Tween 20), Lysis Buffer 10x, Substrate Solution (tetramethyl-benzidine), Stop Solution (1M Phosphoric acid), Adhesive Films
96 Tests
Cell lysate
Storage of components varies. See protocol for specific instructions.
Apoptosis, Cellular Markers, Cholesterol Metabolism, Core ESC Like Genes, Lipid and Metabolism, Mitochondrial Markers, Stem Cell Markers
CYCHCS, cytochrome c, cytochrome c, somatic, THC4