Learn More
Raybiotech IncSupplier Diversity Partner Human Cytokeratin 8/KRT8 ELISA Kit DFS Item
Manufacturer:  Raybiotech IncSupplier Diversity Partner ELHKRT81
For Serum, Plasma, and Cell Culture Supernatants. Sensitivity: 0.4 ng/ml. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: This ELISA antibody pair recognizes Human Cytokeratin 8.
Catalog No. 50-194-9483