Learn More
Raybiotech IncSupplier Diversity Partner Quantibody Human Cytokine Array 1 Kit. DFS Item
Manufacturer:  Raybiotech IncSupplier Diversity Partner QAHCYT11
Detects 20 Human Cytokines. Suitable for all liquid sample types. Quantitative, Sandwich-based, Glass Slide Antibody Array. Simultaneous quantitative measurement of multiple proteins in a wide variety of sample types. Quantibody combines the high specificity and sensitivity of ELISA with the high throughput of the glass chip-based array. Less sample, more data: only 50 ul of sample is needed for quantification of up to 40 cytokines. Proteins detected: GM-CSF, GRO alpha/beta/gamma, IFN-gamma, IL-1 alpha (IL-1 F1), IL-1 beta (IL-1 F2), IL-10, IL-12 p70, IL-13, IL-2, IL-4, IL-5, IL-6, IL-8 (CXCL8), MCP-1 (CCL2), MIP-1 alpha (CCL3), MIP-1 beta (CCL4), MMP-9, RANTES (CCL5), TNF alpha, VEGF-A
Catalog No. 50-195-0815