Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Cytosol Nonspecific Dipeptidase (CNDP2)/CPGL Antibody, R&D Systems™

Sheep Polyclonal Antibody
$200.50 - $526.50
Specifications
Antigen | Cytosol Nonspecific Dipeptidase (CNDP2)/CPGL |
---|---|
Concentration | LYOPH |
Dilution | Western Blot 0.1 ug/mL, Immunoprecipitation 25 ug/mL |
Applications | Western Blot, Immunoprecipitation |
Classification | Polyclonal |
Description
Cytosol Nonspecific Dipeptidase (CNDP2)/CPGL Polyclonal specifically detects Cytosol Nonspecific Dipeptidase (CNDP2)/CPGL in Human samples. It is validated for Western Blot, Immunoprecipitation.Specifications
Cytosol Nonspecific Dipeptidase (CNDP2)/CPGL | |
Western Blot 0.1 ug/mL, Immunoprecipitation 25 ug/mL | |
Polyclonal | |
Sheep | |
Human | |
CN2, CN2cytosolic non-specific dipeptidase, CNDP dipeptidase 2, CNDP dipeptidase 2 (metallopeptidase M20 family), CNDP2, CPGL, CPGLcytosolic nonspecific dipeptidase, FLJ10830, HsT2298, PEPA, PEPAEC 3.4.13.18, Peptidase AGlutamate carboxypeptidase-like protein 1 | |
CNDP2 | |
IgG | |
Affinity Purified | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
LYOPH | |
Western Blot, Immunoprecipitation | |
Unconjugated | |
RUO | |
Q96KP4 | |
55748 | |
S. frugiperda insect ovarian cell line Sf 21-derived recombinant human Cytosol Nonspecific Dipeptidase (CNDP2)/CPGL Ala2-Asp475 Accession # Q96KP4 | |
Primary | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Detects human Cytosol Nonspecific Dipeptidase (CNDP2)/CPGL in direct ELISAs and Western blots. In direct ELISAs and Western blots, less than 1% cross-reactivity with recombinant human CPGL-2 is observed. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title