Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human DAB2 Full-length ORF (AAH03064, 1 a.a. - 770 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Manufacturer:  Abnova™ H00001601P01L

 View more versions of this product

Catalog No. 89-941-380


Add to cart

Description

Description

DAB2 mRNA is expressed in normal ovarian epithelial cells but is down-regulated or absent from ovarian carcinoma cell lines. The 770-amino acid predicted protein has an overall 83% identity with the mouse p96 protein, a putative mitogen-responsive phosphoprotein; homology is strongest in the amino-terminal end of the protein in a region corresponding to the phosphotyrosine interaction domain. The down-regulation of DAB2 may play an important role in ovarian carcinogenesis. This gene was initially named DOC2 (for Differentially expressed in Ovarian Cancer) and is distinct from the DOC2A and DOC2B genes (for double C2-like domains, alpha and beta). [provided by RefSeq]

Sequence: MSNEVETSATNGQPDQQAAPKAPSKKEKKKGPEKTDEYLLARFKGDGVKYKAKLIGIDDVPDARGDKMSQDSMMKLKGMAAAGRSQGQHKQRIWVNISLSGIKIIDEKTGVIEHEHPVNKISFIARDVTDNRAFGYVCGGEGQHQFFAIKTGQQAEPLVVDLKDLFQVIYNVKKKEEEKKKIEEASKAVENGSEALMILDDQTNKLKSGVDQMDLFGDMSTPPDLNSPTESKDILLVDLNSEIDTNQNSLRENPFLTNGITSCSLPRPTPQASFLPENAFSANLNFFPTPNPDPFRDDPFTQPDQSTPSSFDSLKSPDQKKENSSSSSTPLSNGPLNGDVDYFGQQFDQISNRTGKQEAQAGPWPFSSSQTQPAVRTQNGVSEREQNGFSVKSSPNPFVGSPPKGLSIQNGVKQDLESSVQSSPHDSIAIIPPPQSTKPGRGRRTAKSSANDLLASDIFAPPVSEPSGQASPTGQPTALQPNPLDLFKTSAPAPVGPLVGLGGVTVTLPQAGPWNTASLVFNQSPSMAPGAMMGGQPSGFSQPVIFGTSPAVSGWNQPSPFAASTPPPVPVVWGPSASVAPNAWSTTSPLGNPFQSNIFPAPAVSTQPPSMHSSLLVTPPQPPPRAGPPKDISSDAFTALDPLGDKEIKDVKEMFKDFQLWQPPAVPARKGEQTSSGTLSAFASYFNSKVGIPQENADHDDFDANQLLNKINEPPKPAPRQVSLPVTKSTDNAFENPFFKDSFGSSQASVASSQPVSSEMYRDPFGNPFA
Specifications

Specifications

AAH03064
Liquid
1601
DAB2 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MSNEVETSATNGQPDQQAAPKAPSKKEKKKGPEKTDEYLLARFKGDGVKYKAKLIGIDDVPDARGDKMSQDSMMKLKGMAAAGRSQGQHKQRIWVNISLSGIKIIDEKTGVIEHEHPVNKISFIARDVTDNRAFGYVCGGEGQHQFFAIKTGQQAEPLVVDLKDLFQVIYNVKKKEEEKKKIEEASKAVENGSEALMILDDQTNKLKSGVDQMDLFGDMSTPPDLNSPTESKDILLVDLNSEIDTNQNSLRENPFLTNGITSCSLPRPTPQASFLPENAFSANLNFFPTPNPDPFRDDPFTQPDQSTPSSFDSLKSPDQKKENSSSSSTPLSNGPLNGDVDYFGQQFDQISNRTGKQEAQAGPWPFSSSQTQPAVRTQNGVSEREQNGFSVKSSPNPFVGSPPKGLSIQNGVKQDLESSVQSSPHDSIAIIPPPQSTKPGRGRRTAKSSANDLLASDIFAPPVSEPSGQASPTGQPTALQPNPLDLFKTSAPAPVGPLVGLGGVTVTLPQAGPWNTASLVFNQSPSMAPGAMMGGQPSGFSQPVIFGTSPAVSGWNQPSPFAASTPPPVPVVWGPSASVAPNAWSTTSPLGNPFQSNIFPAPAVSTQPPSMHSSLLVTPPQPPPRAGPPKDISSDAFTALDPLGDKEIKDVKEMFKDFQLWQPPAVPARKGEQTSSGTLSAFASYFNSKVGIPQENADHDDFDANQLLNKINEPPKPAPRQVSLPVTKSTDNAFENPFFKDSFGSSQASVASSQPVSSEMYRDPFGNPFA
RUO
DAB2
wheat germ expression system
Yes
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
110.22kDa
Glutathione Sepharose 4 Fast Flow
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
DOC-2|DOC2|FLJ26626
DAB2
Wheat Germ (in vitro)
GST
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit