Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human DALRD3 Full-length ORF (NP_001009996.1, 1 a.a. - 543 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Supplier:  Abnova™ H00055152P01S

 View more versions of this product

Catalog No. 89-941-398


Add to Cart

Description

Description

Sequence: MATRRLGVGETLGALNAALGPGGPVWIKETRTRHLRSRDFLAPHRALQARFDDGQVPEHLLHALACLQGPGVAPVLRCAPTPAGLSLQLQRSAVFERVLSAVAAYATPASPASLGQRVLLHCPALRSSPCALRLSQLRTVLVADHLARALRAHGVCVRLVPAVRDPHMLTFLQQLRVDWPAASERASSHTLRSHALEELTSANDGRTLSPGILGRLCLKELVEEQGRTAGYDPNLDNCLVTEDLLSVLAELQEALWHWPEDSHPGLAGASDTGTGGCLVVHVVSCEEEFQQQKLDLLWQKLVDKAPLRQKHLICGPVKVAGAPGTLMTAPEYYEFRHTQVCKASALKHGGDLAQDPAWTEIFGVLSVATIKFEMLSTAPQSQLFLALADSSISTKGTKSGTFVMYNCARLATLFESYKCSMEQGLYPTFPPVSSLDFSLLHDEGEWLLLFNSILPFPDLLSRTAVLDCTAPGLHIAVRTEMICKFLVQLSMDFSSYYNRVHILGEPRPHLFGQMFVRLQLLRAVREVLHTGLAMLGLPPLSHI
Specifications

Specifications

NP_001009996.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
85.80kDa
Glutathione Sepharose 4 Fast Flow
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
FLJ10496
DALRD3
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
55152
DALRD3 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MATRRLGVGETLGALNAALGPGGPVWIKETRTRHLRSRDFLAPHRALQARFDDGQVPEHLLHALACLQGPGVAPVLRCAPTPAGLSLQLQRSAVFERVLSAVAAYATPASPASLGQRVLLHCPALRSSPCALRLSQLRTVLVADHLARALRAHGVCVRLVPAVRDPHMLTFLQQLRVDWPAASERASSHTLRSHALEELTSANDGRTLSPGILGRLCLKELVEEQGRTAGYDPNLDNCLVTEDLLSVLAELQEALWHWPEDSHPGLAGASDTGTGGCLVVHVVSCEEEFQQQKLDLLWQKLVDKAPLRQKHLICGPVKVAGAPGTLMTAPEYYEFRHTQVCKASALKHGGDLAQDPAWTEIFGVLSVATIKFEMLSTAPQSQLFLALADSSISTKGTKSGTFVMYNCARLATLFESYKCSMEQGLYPTFPPVSSLDFSLLHDEGEWLLLFNSILPFPDLLSRTAVLDCTAPGLHIAVRTEMICKFLVQLSMDFSSYYNRVHILGEPRPHLFGQMFVRLQLLRAVREVLHTGLAMLGLPPLSHI
RUO
DALRD3
Wheat Germ (in vitro)
GST
Liquid
Product Suggestions

Product Suggestions

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit