Learn More
Abnova™ Human DAND5 Partial ORF (NP_689867, 90 a.a. - 188 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Supplier: Abnova™ H00199699Q01S
Description
This gene encodes a member of the BMP (bone morphogenic protein) antagonist family. Like BMPs, BMP antagonists contain cystine knots and typically form homo- and heterodimers. The CAN (cerberus and dan) subfamily of BMP antagonists, to which this gene belongs, is characterized by a C-terminal cystine knot with an eight-membered ring. The antagonistic effect of the secreted protein encoded by this gene is likely due to its direct binding to BMP proteins. As an antagonist of BMP, this gene may play a role in regulating organogenesis, body patterning, and tissue differentiation. In mouse, this protein has been shown to bind Nodal and to inhibit the Nodal signaling pathway which patterns left/right body asymmetry. [provided by RefSeq]
Sequence: PLNPQEVIQGMCKAVPFVQVFSRPGCSAIRLRNHLCFGHCSSLYIPGSDPTPLVLCNSCMPARKRWAPVVLWCLTGSSASRRRVKISTMLIEGCHCSPKSpecifications
NP_689867 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.63kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
PLNPQEVIQGMCKAVPFVQVFSRPGCSAIRLRNHLCFGHCSSLYIPGSDPTPLVLCNSCMPARKRWAPVVLWCLTGSSASRRRVKISTMLIEGCHCSPK | |
RUO | |
DAND5 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
199699 | |
DAND5 (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
CER2/CERL2/CKTSF1B3/COCO/CRL2/DANTE/GREM3/MGC126849/SP1 | |
DAND5 | |
Recombinant | |
wheat germ expression system |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.