Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human DAPK1 (NP_004929.1, 1 a.a. - 289 a.a.) Partial Recombinant Protein with His tag expressed in Baculovirus infected Sf21 cells

Used for Func, SDS-PAGE

Manufacturer:  Abnova™ P5696

Catalog No. 89-941-413


Add to cart

Description

Description

Death-associated protein kinase 1 is a positive mediator of gamma-interferon induced programmed cell death. DAPK1 encodes a structurally unique 160-kD calmodulin dependent serine-threonine kinase that carries 8 ankyrin repeats and 2 putative P-loop consensus sites. It is a tumor suppressor candidate. [provided by RefSeq]

Specifications

Specifications

NP_004929.1
Liquid
1612
DAPK1 (Human) Recombinant Protein
Ni-NTA affinity chromatography
Loading 1 ug protein in SDS-PAGE
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
DAPK|DKFZp781I035
DAPK1
The activity was measured by off-chip mobility shift assay. The enzyme was incubated with fluorescence-labeled substrate and Mg (or Mn)/ATP. The phosphorylated and unphosphorylated substrates were separated and detected by LabChip™3000.
Substrate : DAPK1tide. ATP: 100 µM.
Yes
Functional Study, Sodium Dodecyl Sulfate–Polyacrylamide Gel Electrophoresis
Liquid
60kDa
Baculovirus expression system
94% by SDS-PAGE/CBB staining.
5 ug
RUO
DAPK1
Baculovirus expression system
Insect
His
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit