Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Abnova™ Human DAPK3 (NP_001339.1, 1 a.a. - 454 a.a.) Full-length Recombinant Protein with GST-tag expressed in baculovirus infected Sf21 cells
Used for Func, SDS-PAGE
Supplier: Abnova™ P5531
Description
Death-associated protein kinase 3 (DAPK3) induces morphological changes in apoptosis when overexpressed in mammalian cells. These results suggest that DAPK3 may play a role in the induction of apoptosis. [provided by RefSeq]
Specifications
NP_001339.1 | |
Liquid | |
80kDa | |
Baculovirus infected insect cell (Sf21) expression system | |
Loading 1 ug protein in SDS-PAGE | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
FLJ36473/ZIP/ZIPK | |
DAPK3 | |
Baculovirus | |
GST | |
Liquid |
Functional Study, SDS-PAGE | |
1613 | |
DAPK3 (Human) Recombinant Protein | |
Glutathione sepharose chromatography | |
5 ug | |
RUO | |
DAPK3 | |
The activity was measured by off-chip mobility shift assay. The enzyme was incubated with fluorescence-labeled substrate and Mg(or Mn)/ATP. The phosphorylated and unphosphorylated substrates were separated and detected by LabChip 3000. Substrate: ZIPtide. ATP: 100 uM. | |
Recombinant | |
Baculovirus infected insect cell (Sf21) expression system | |
67% by SDS-PAGE/CBB staining |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction