Learn More
Abnova™ Human DCLK1 (NP_004725.1, 1 a.a. - 729 a.a.) Full-length Recombinant Protein with GST-tag expressed in baculovirus infected Sf21 cells
Used for Func, SDS-PAGE
Manufacturer:  Abnova™ P5532
Catalog No. 89-941-509
Add to cart
Description
Description
DCAMKL1 is a microtubule-associated protein that phosphorylates itself and myelin basic protein (MBP; MIM 159430). DCAMKL1 has microtubule polymerizing activity that is independent of its protein kinase activity (Lin et al., 2000 [PubMed 11124993]).[supplied by OMIM]
Specifications
Specifications
NP_004725.1
Liquid
9201
DCLK1 (Human) Recombinant Protein
Glutathione sepharose chromatography
Loading 1 ug protein in SDS-PAGE
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
DCAMKL1|DCDC3A|DCLK|KIAA0369
DCLK1
The activity was measured by off-chip mobility shift assay. The enzyme was incubated with fluorescence-labeled substrate and Mg(or Mn)/ATP. The phosphorylated and unphosphorylated substrates were separated and detected by LabChip 3000. Substrate: GS peptide. ATP: 1000 uM
Yes
Functional Study, Sodium Dodecyl Sulfate–Polyacrylamide Gel Electrophoresis
Liquid
108kDa
Baculovirus infected insect cell (Sf21) expression system
58% by SDS-PAGE/CBB staining
5 ug
RUO
DCLK1
Baculovirus infected insect cell (Sf21) expression system
Insect
GST