Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human DCLRE1B Full-length ORF (NP_073747.1, 1 a.a. - 532 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Manufacturer:  Abnova™ H00064858P01

Catalog No. 89-941-511


Add to cart

Description

Description

DNA interstrand cross-links prevent strand separation, thereby physically blocking transcription, replication, and segregation of DNA. DCLRE1B is one of several evolutionarily conserved genes involved in repair of interstrand cross-links (Dronkert et al., 2000 [PubMed 10848582]).[supplied by OMIM]

Sequence: MNGVLIPHTPIAVDFWSLRRAGTARLFFLSHMHSDHTVGLSSTWARPLYCSPITAHLLHRHLQVSKQWIQALEVGESHVLPLDEIGQETMTVTLLDANHCPGSVMFLFEGYFGTILYTGDFRYTPSMLKEPALTLGKQIHTLYLDNTNCNPALVLPSRQEAAHQIVQLIRKHPQHNIKIGLYSLGKESLLEQLALEFQTWVVLSPRRLELVQLLGLADVFTVEEKAGRIHAVDHMEICHSNMLRWNQTHPTIAILPTSRKIHSSHPDIHVIPYSDHSSYSELRAFVAALKPCQVVPIVSRRPCGGFQDSLSPRISVPLIPDSVQQYMSSSSRKPSLLWLLERRLKRPRTQGVVFESPEESADQSQADRDSKKAKKEKLSPWPADLEKQPSHHPLRIKKQLFPDLYSKEWNKAVPFCESQKRVTMLTAPLGFSVHLRSTDEEFISQKTREEIGLGSPLVPMGDDDGGPEATGNQSAWMGHGSPLSHSSKGTPLLATEFRGLALKYLLTPVNFFQAGYSSRRFDQQVEKYHKPC
Specifications

Specifications

NP_073747.1
Liquid
64858
DCLRE1B (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MNGVLIPHTPIAVDFWSLRRAGTARLFFLSHMHSDHTVGLSSTWARPLYCSPITAHLLHRHLQVSKQWIQALEVGESHVLPLDEIGQETMTVTLLDANHCPGSVMFLFEGYFGTILYTGDFRYTPSMLKEPALTLGKQIHTLYLDNTNCNPALVLPSRQEAAHQIVQLIRKHPQHNIKIGLYSLGKESLLEQLALEFQTWVVLSPRRLELVQLLGLADVFTVEEKAGRIHAVDHMEICHSNMLRWNQTHPTIAILPTSRKIHSSHPDIHVIPYSDHSSYSELRAFVAALKPCQVVPIVSRRPCGGFQDSLSPRISVPLIPDSVQQYMSSSSRKPSLLWLLERRLKRPRTQGVVFESPEESADQSQADRDSKKAKKEKLSPWPADLEKQPSHHPLRIKKQLFPDLYSKEWNKAVPFCESQKRVTMLTAPLGFSVHLRSTDEEFISQKTREEIGLGSPLVPMGDDDGGPEATGNQSAWMGHGSPLSHSSKGTPLLATEFRGLALKYLLTPVNFFQAGYSSRRFDQQVEKYHKPC
RUO
DCLRE1B
wheat germ expression system
Yes
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
86.4kDa
Glutathione Sepharose 4 Fast Flow
2 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
APOLLO|DKFZp686C24187|FLJ12810|FLJ13998|SNM1B
DCLRE1B
Wheat Germ (in vitro)
GST
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit