Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human DCLRE1C Full-length ORF (AAH22254, 1 a.a. - 434 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Manufacturer:  Abnova™ H00064421P01

Catalog No. 89-941-514


Add to cart

Description

Description

This gene encodes a nuclear protein that is involved in V(D)J recombination and DNA repair. The protein has single-strand-specific 5'-3' exonuclease activity; it also exhibits endonuclease activity on 5' and 3' overhangs and hairpins when complexed with protein kinase, DNA-activated, catalytic polypeptide. Mutations in this gene cause Athabascan-type severe combined immunodeficiency (SCIDA). [provided by RefSeq]

Sequence: MSSFEGQMAEYPTISIDRFDRENLRARAYFLSHCHKDHMKGLRAPTLKRRLECSLKVYLYCSPVTKELLLTSPKYRFWKKRIISIEIETPTQISLVDEASGEKEEIVVTLLPAGHCPGSVMFLFQGNNGTVLYTGDFRLAQGEAARMELLHSGGRVKDIQSVYLDTTFCDPRFYQIPSREECLSGVLELVRSWITRSPYHVVWLNCKAAYGYEYLFTNLSEELGVQVHVNKLDMFRNMPEILRHLTTDRNTQIHACRHPKAEEYFQWSKLPCGITSRNRIPLHIISIKPSTMWFGERSRKTNVIVRTGESSYRACFSFHSSYSEIKDFLSYLCPVNAYPNVIPVGTTMDKVVEILKPLCRSSQSTEPKYKPLGKLKRARTVHRDSGSHSVTQARMRWCHHDSLYPLTPGIKRSSCLSLLTSWITGAYRHAQLMI
Specifications

Specifications

AAH22254
Liquid
64421
DCLRE1C (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MSSFEGQMAEYPTISIDRFDRENLRARAYFLSHCHKDHMKGLRAPTLKRRLECSLKVYLYCSPVTKELLLTSPKYRFWKKRIISIEIETPTQISLVDEASGEKEEIVVTLLPAGHCPGSVMFLFQGNNGTVLYTGDFRLAQGEAARMELLHSGGRVKDIQSVYLDTTFCDPRFYQIPSREECLSGVLELVRSWITRSPYHVVWLNCKAAYGYEYLFTNLSEELGVQVHVNKLDMFRNMPEILRHLTTDRNTQIHACRHPKAEEYFQWSKLPCGITSRNRIPLHIISIKPSTMWFGERSRKTNVIVRTGESSYRACFSFHSSYSEIKDFLSYLCPVNAYPNVIPVGTTMDKVVEILKPLCRSSQSTEPKYKPLGKLKRARTVHRDSGSHSVTQARMRWCHHDSLYPLTPGIKRSSCLSLLTSWITGAYRHAQLMI
RUO
DCLRE1C
wheat germ expression system
Yes
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
73.48kDa
Glutathione Sepharose 4 Fast Flow
2 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
A-SCID|DCLREC1C|FLJ11360|FLJ36438|RS-SCID|SCIDA|SNM1C
DCLRE1C
Wheat Germ (in vitro)
GST
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit