Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human DCTN2 Full-length ORF (AAH00718.1, 1 a.a. - 401 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Manufacturer:  Abnova™ H00010540P01L

 View more versions of this product

Catalog No. 89-941-532


Add to cart

Description

Description

This gene encodes a 50-kD subunit of dynactin, a macromolecular complex consisting of 10-11 subunits ranging in size from 22 to 150 kD. Dynactin binds to both microtubules and cytoplasmic dynein. It is involved in a diverse array of cellular functions, including ER-to-Golgi transport, the centripetal movement of lysosomes and endosomes, spindle formation, chromosome movement, nuclear positioning, and axonogenesis. This subunit is present in 4-5 copies per dynactin molecule. It contains three short alpha-helical coiled-coil domains that may mediate association with self or other dynactin subunits. It may interact directly with the largest subunit (p150) of dynactin and may affix p150 in place. [provided by RefSeq]

Sequence: MADPKYADLPGIARNEPDVYETSDLPEDDQAEFDAEELTSTSVEHIIVNPNAAYDKFKDKRVGTKGLDFSDRIGKTKRTGYESGEYEMLGEGLGVKETPQQKYQRLLHEVQELTTEVEKIKTTVKESATEEKLTPVLLAKQLAALKQQLVASHLEKLLGPDAAINLTDPDGALAKRLLLQLEATKNSKGGSGGKTTGTPPDSSLVTYELHSRPEQDKFSQAAKVAELEKRLTELETAVRCDQDAQNPLSAGLQGACLMETVELLQAKVSALDLAVLDQVEARLQSVLGKVNEIAKHKASVEDADTQSKVHQLYETIQRWSPIASTLPELVQRLVTIKQLHEQAMQFGQLLTHLDTTQQMIANSLKDNTTLLTQVQTTMRENLATVEGNFASIDERMKKLGK
Specifications

Specifications

AAH00718.1
Liquid
10540
DCTN2 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MADPKYADLPGIARNEPDVYETSDLPEDDQAEFDAEELTSTSVEHIIVNPNAAYDKFKDKRVGTKGLDFSDRIGKTKRTGYESGEYEMLGEGLGVKETPQQKYQRLLHEVQELTTEVEKIKTTVKESATEEKLTPVLLAKQLAALKQQLVASHLEKLLGPDAAINLTDPDGALAKRLLLQLEATKNSKGGSGGKTTGTPPDSSLVTYELHSRPEQDKFSQAAKVAELEKRLTELETAVRCDQDAQNPLSAGLQGACLMETVELLQAKVSALDLAVLDQVEARLQSVLGKVNEIAKHKASVEDADTQSKVHQLYETIQRWSPIASTLPELVQRLVTIKQLHEQAMQFGQLLTHLDTTQQMIANSLKDNTTLLTQVQTTMRENLATVEGNFASIDERMKKLGK
RUO
DCTN2
wheat germ expression system
Yes
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
70.6kDa
Glutathione Sepharose 4 Fast Flow
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
DCTN50|DYNAMITIN|RBP50
DCTN2
Wheat Germ (in vitro)
GST
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit