Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human DDEF2 Full-length ORF (NP_003878.1, 1 a.a. - 1006 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Manufacturer:  Abnova™ H00008853P01S

 View more versions of this product

Catalog No. 89-941-577


Add to cart

Description

Description

This gene encodes a multidomain protein containing an N-terminal alpha-helical region with a coiled-coil motif, followed by a pleckstrin homology (PH) domain, an Arf-GAP domain, an ankyrin homology region, a proline-rich region, and a C-terminal Src homology 3 (SH3) domain. The protein localizes in the Golgi apparatus and at the plasma membrane, where it colocalizes with protein tyrosine kinase 2-beta (PYK2). The encoded protein forms a stable complex with PYK2 in vivo. This interaction appears to be mediated by binding of its SH3 domain to the C-terminal proline-rich domain of PYK2. The encoded protein is tyrosine phosphorylated by activated PYK2. It has catalytic activity for class I and II ArfGAPs in vitro, and can bind the class III Arf ARF6 without immediate GAP activity. The encoded protein is believed to function as an ARF GAP that controls ARF-mediated vesicle budding when recruited to Golgi membranes. In addition, it functions as a substrate and downstream target for PYK2 and SRC, a pathway that may be involved in the regulation of vesicular transport. Multiple transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq]

Sequence: MPDQISVSEFVAETHEDYKAPTASSFTTRTAQCRNTVAAIEEALDVDRMVLYKMKKSVKAINSSGLAHVENEEQYTQALEKFGGNCVCRDDPDLGSAFLKFSVFTKELTALFKNLIQNMNNIISFPLDSLLKGDLKGVKGDLKKPFDKAWKDYETKITKIEKEKKEHAKLHGMIRTEISGAEIAEEMEKERRFFQLQMCEYLLKVNEIKIKKGVDLLQNLIKYFHAQCNFFQDGLKAVESLKPSIETLSTDLHTIKQAQDEERRQLIQLRDILKSALQVEQKEDSQIRQSTAYSLHQPQGNKEHGTERNGSLYKKSDGIRKVWQKRKCSVKNGFLTISHGTANRPPAKLNLLTCQVKTNPEEKKCFDLISHDRTYHFQAEDEQECQIWMSVLQNSKEEALNNAFKGDDNTGENNIVQELTKEIISEVQRMTGNDVCCDCGAPDPTWLSTNLGILTCIECSGIHRELGVHYSRMQSLTLDVLGTSELLLAKNIGNAGFNEIMECCLPAEDSVKPNPGSDMNARKDYITAKYIERRYARKKHADNAAKLHSLCEAVKTRDIFGLLQAYADGVDLTEKIPLANGHEPDETALHLAVRSVDRTSLHIVDFLVQNSGNLDKQTGKGSTALHYCCLTDNAECLKLLLRGKASIEIANESGETPLDIAKRLKHEHCEELLTQALSGRFNSHVHVEYEWRLLHEDLDESDDDMDEKLQPSPNRREDRPISFYQLGSNQLQSNAVSLARDAANLAKEKQRAFMPSILQNETYGALLSGSPPPAQPAAPSTTSAPPLPPRNVGKVQTASSANTLWKTNSVSVDGGSRQRSSSDPPAVHPPLPPLRVTSTNPLTPTPPPPVAKTPSVMEALSQPSKPAPPGISQIRPPPLPPQPPSRLPQKKPAPGADKSTPLTNKGQPRGPVDLSATEALGPLSNAMVLQPPAPMPRKSQATKLKPKRVKALYNCVADNPDELTFSEGDVIIVDGEEDQEWWIGHIDGDPGRKGAFPVSFVHFIAD
Specifications

Specifications

NP_003878.1
Liquid
8853
DDEF2 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
AMAP2|CENTB3|DDEF2|FLJ42910|KIAA0400|PAG3|PAP|Pap-alpha|SHAG1
ASAP2
Wheat Germ (in vitro)
GST
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
138.1kDa
Glutathione Sepharose 4 Fast Flow
10 ug
RUO
ASAP2
wheat germ expression system
Yes
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit