Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human DDEFL1 Full-length ORF (NP_060177.2, 1 a.a. - 903 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Manufacturer:  Abnova™ H00055616P01S

 View more versions of this product

Catalog No. 89-941-581


Add to cart

Description

Description

This gene encodes a member of a subfamily of ADP-ribosylation factor(Arf) GTPase-activating proteins that contain additional ankyrin repeat and pleckstrin homology domains. The Arf GAP domain of this protein catalyzes the hydrolysis of GTP bound to Arf proteins. The encoded protein promotes cell differentiation and migration and has been implicated in cancer cell invasion. Alternative splicing results in multiple transcript variants. [provided by RefSeq]

Sequence: MPEQFSVAEFLAVTAEDLSSPAGAAAFAAKMPRYRGAALAREEILEGDQAILQRIKKAVRAIHSSGLGHVENEEQYREAVESLGNSHLSQNSHELSTGFLNLAVFTREVAALFKNLIQNLNNIVSFPLDSLMKGQLRDGRQDSKKQLEKAWKDYEAKMAKLEKERDRARVTGGIPGEVAQDMQRERRIFQLHMCEYLLKAGESQMKQGPDFLQSLIKFFHAQHNFFQDGWKAAQSLFPFIEKLAASVHALHQAQEDELQKLTQLRDSLRGTLQLESREEHLSRKNSGCGYSIHQHQGNKQFGTEKVGFLYKKSDGIRRVWQKRKCGVKYGCLTISHSTINRPPVKLTLLTCQVRPNPEEKKCFDLVTHNRTYHFQAEDEHECEAWVSVLQNSKDEALSSAFLGEPSAGPGSWGSAGHDGEPHDLTKLLIAEVKSRPGNSQCCDCGAADPTWLSTNLGVLTCIQCSGVHRELGVRFSRMQSLTLDLLGPSELLLALNMGNTSFNEVMEAQLPSHGGPKPSAESDMGTRRDYIMAKYVEHRFARRCTPEPQRLWTAICNRDLLSVLEAFANGQDFGQPLPGPDAQAPEELVLHLAVKVANQASLPLVDFIIQNGGHLDAKAADGNTALHYAALYNQPDCLKLLLKGRALVGTVNEAGETALDIARKKHHKECEELLEQAQAGTFAFPLHVDYSWVISTEPGSDSEEDEEEKRCLLKLPAQAHWASGRLDISNKTYETVASLGAATPQGESEDCPPPLPVKNSSRTLVQGCARHASGDRSEVSSLSSEAPETPESLGSPASSSSLMSPLEPGDPSQAPPNSEEGLREPPGTSRPSLTSGTTPSEMYLPVRFSSESTRSYRRGARSPEDGPSARQPLPRRNVPVGITEGDGSRTGSLPASSVQLLQD
Specifications

Specifications

wheat germ expression system
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
125.6kDa
Glutathione Sepharose 4 Fast Flow
10 ug
RUO
ASAP3
Wheat Germ (in vitro)
GST
NP_060177.2
Liquid
55616
DDEFL1 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
ACAP4/CENTB6/DDEFL1/FLJ20199/UPLC1
ASAP3
Yes
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit