Learn More
Abnova™ Human DDX17 Full-length ORF (NP_112020.1, 1 a.a. - 183 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Supplier: Abnova™ H00010521P01L
Description
DEAD box proteins, characterized by the conserved motif Asp-Glu-Ala-Asp (DEAD), are putative RNA helicases. They are implicated in a number of cellular processes involving alteration of RNA secondary structure, such as translation initiation, nuclear and mitochondrial splicing, and ribosome and splicesosome assembly. Based on their distribution patterns, some members of this family are believed to be involved in embryogenesis, spermatogenesis, and cellular growth and division. This gene encodes a DEAD box protein, which is an ATPase activated by a variety of RNA species, but not by dsDNA. This protein, and that encoded by DDX5 gene, are more closely related to each other than to any other member of the DEAD box family. Several alternatively spliced transcripts encoding different isoforms, some of which use non-AUG (CUG) translation initiation codon, have been described for this gene. [provided by RefSeq]
Sequence: MQLVDHRGGGGGGGKGGRSRYRTTSSANNPNLMYQDECDRRLRGVKDGGRRDSASYRDRSETDRAGYANGSGYGSPNSAFGAQAGQYTYGQGTYGAAAYGTSSYTAQEYGAGTYGASSTTSTGRSSQSSSQQFSGIGRSGQQPQPLMSQQFAQPPGATNMIGYMGQTAYQYPPPPPPPPPSRKSpecifications
NP_112020.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
45.6kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
DKFZp761H2016/P72/RH70 | |
DDX17 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
10521 | |
DDX17 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MQLVDHRGGGGGGGKGGRSRYRTTSSANNPNLMYQDECDRRLRGVKDGGRRDSASYRDRSETDRAGYANGSGYGSPNSAFGAQAGQYTYGQGTYGAAAYGTSSYTAQEYGAGTYGASSTTSTGRSSQSSSQQFSGIGRSGQQPQPLMSQQFAQPPGATNMIGYMGQTAYQYPPPPPPPPPSRK | |
RUO | |
DDX17 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.