Learn More
Abnova™ Human DDX20 Partial ORF (NP_009135, 725 a.a. - 824 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Supplier: Abnova™ H00011218Q01S
Description
DEAD box proteins, characterized by the conserved motif Asp-Glu-Ala-Asp (DEAD), are putative RNA helicases. They are implicated in a number of cellular processes involving alteration of RNA secondary structure such as translation initiation, nuclear and mitochondrial splicing, and ribosome and spliceosome assembly. Based on their distribution patterns, some members of this family are believed to be involved in embryogenesis, spermatogenesis, and cellular growth and division. This gene encodes a DEAD box protein, which has an ATPase activity and is a component of the survival of motor neurons (SMN) complex. This protein interacts directly with SMN, the spinal muscular atrophy gene product, and may play a catalytic role in the function of the SMN complex on RNPs. [provided by RefSeq]
Sequence: EGASQRAKQSRRNLPRRSSFRLQTEAQEDDWYDCHREIRLSFSDTYQDYEEYWRAYYRAWQEYYAAASHSYYWNAQRHPSWMAAYHMNTIYLQEMMHSNQSpecifications
NP_009135 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.74kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
EGASQRAKQSRRNLPRRSSFRLQTEAQEDDWYDCHREIRLSFSDTYQDYEEYWRAYYRAWQEYYAAASHSYYWNAQRHPSWMAAYHMNTIYLQEMMHSNQ | |
RUO | |
DDX20 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
11218 | |
DDX20 (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
DKFZp434H052/DP103/GEMIN3 | |
DDX20 | |
Recombinant | |
wheat germ expression system |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.