Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human DDX28 Full-length ORF (NP_060850.1, 1 a.a. - 540 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Manufacturer:  Abnova™ H00055794P01

Catalog No. 89-941-665


Add to cart

Description

Description

DEAD box proteins, characterized by the conserved motif Asp-Glu-Ala-Asp (DEAD), are putative RNA helicases. They are implicated in a number of cellular processes involving alteration of RNA secondary structure, such as translation initiation, nuclear and mitochondrial splicing, and ribosome and spliceosome assembly. Based on their distribution patterns, some members of the DEAD box protein family are believed to be involved in embryogenesis, spermatogenesis, and cellular growth and division. This gene is intronless. It encodes an RNA-dependent ATPase. The encoded protein is localized in the mitochondria and the nucleus, and can be transported between the mitochondria and the nucleus. [provided by RefSeq]

Sequence: MALARPVRLFSLVTRLLLAPRRGLTVRSPDEPLPVVRIPVALQRQLEQRQSRRRNLPRPVLVRPGPLLVSARRPELNQPARLTLGRWERAPLASQGWKSRRARRDHFSIERAQQEAPAVRKLSSKGSFADLGLEPRVLHALQEAAPEVVQPTTVQSSTIPSLLRGRHVVCAAETGSGKTLSYLLPLLQRLLGQPSLDSLPIPAPRGLVLVPSRELAQQVRAVAQPLGRSLGLLVRDLEGGHGMRRIRLQLSRQPSADVLVATPGALWKALKSRLISLEQLSFLVLDEADTLLDESFLELVDYILEKSHIAEGPADLEDPFNPKAQLVLVGATFPEGVGQLLNKVASPDAVTTITSSKLHCIMPHVKQTFLRLKGADKVAELVHILKHRDRAERTGPSGTVLVFCNSSSTVNWLGYILDDHKIQHLRLQGQMPALMRVGIFQSFQKSSRDILLCTDIASRGLDSTGVELVVNYDFPPTLQDYIHRAGRVGRVGSEVPGTVISFVTHPWDVSLVQKIELAARRRRSLPGLASSVKEPLPQAT
Specifications

Specifications

NP_060850.1
Liquid
55794
DDX28 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MALARPVRLFSLVTRLLLAPRRGLTVRSPDEPLPVVRIPVALQRQLEQRQSRRRNLPRPVLVRPGPLLVSARRPELNQPARLTLGRWERAPLASQGWKSRRARRDHFSIERAQQEAPAVRKLSSKGSFADLGLEPRVLHALQEAAPEVVQPTTVQSSTIPSLLRGRHVVCAAETGSGKTLSYLLPLLQRLLGQPSLDSLPIPAPRGLVLVPSRELAQQVRAVAQPLGRSLGLLVRDLEGGHGMRRIRLQLSRQPSADVLVATPGALWKALKSRLISLEQLSFLVLDEADTLLDESFLELVDYILEKSHIAEGPADLEDPFNPKAQLVLVGATFPEGVGQLLNKVASPDAVTTITSSKLHCIMPHVKQTFLRLKGADKVAELVHILKHRDRAERTGPSGTVLVFCNSSSTVNWLGYILDDHKIQHLRLQGQMPALMRVGIFQSFQKSSRDILLCTDIASRGLDSTGVELVVNYDFPPTLQDYIHRAGRVGRVGSEVPGTVISFVTHPWDVSLVQKIELAARRRRSLPGLASSVKEPLPQAT
RUO
DDX28
wheat germ expression system
Yes
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
86kDa
Glutathione Sepharose 4 Fast Flow
2 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
FLJ11282|MDDX28
DDX28
Wheat Germ (in vitro)
GST
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit