Learn More
Abnova™ Human DDX41 Partial ORF (NP_057306, 523 a.a. - 622 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Supplier: Abnova™ H00051428Q01L
Description
DEAD box proteins, characterized by the conserved motif Asp-Glu-Ala-Asp (DEAD), are putative RNA helicases. They are implicated in a number of cellular processes involving alteration of RNA secondary structure, such as translation initiation, nuclear and mitochondrial splicing, and ribosome and spliceosome assembly. Based on their distribution patterns, some members of the DEAD box protein family are believed to be involved in embryogenesis, spermatogenesis, and cellular growth and division. This gene encodes a member of this family. The function of this member has not been determined. Based on studies in Drosophila, the abstrakt gene is widely required during post-transcriptional gene expression. [provided by RefSeq]
Sequence: TGRSGNTGIATTFINKACDESVLMDLKALLLEAKQKVPPVLQVLHCGDESMLDIGGERGCAFCGGLGHRITDCPKLEAMQTKQVSNIGRKDYLAHSSMDFSpecifications
NP_057306 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.74kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
TGRSGNTGIATTFINKACDESVLMDLKALLLEAKQKVPPVLQVLHCGDESMLDIGGERGCAFCGGLGHRITDCPKLEAMQTKQVSNIGRKDYLAHSSMDF | |
RUO | |
DDX41 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
51428 | |
DDX41 (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
ABS/MGC8828 | |
DDX41 | |
Recombinant | |
wheat germ expression system |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.