Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human DDX50 Full-length ORF (NP_076950.1, 1 a.a. - 737 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Manufacturer:  Abnova™ H00079009P01S

 View more versions of this product

Catalog No. 89-941-700


Add to cart

Description

Description

DEAD box proteins, characterized by the conserved motif Asp-Glu-Ala-Asp (DEAD), are putative RNA helicases. They are implicated in a number of cellular processes involving alteration of RNA secondary structure such as translation initiation, nuclear and mitochondrial splicing, and ribosome and spliceosome assembly. Based on their distribution patterns, some members of this DEAD box protein family are believed to be involved in embryogenesis, spermatogenesis, and cellular growth and division. This gene encodes a DEAD box enzyme that may be involved in ribosomal RNA synthesis or processing. This gene and DDX21, also called RH-II/GuA, have similar genomic structures and are in tandem orientation on chromosome 10, suggesting that the two genes arose by gene duplication in evolution. This gene has pseudogenes on chromosomes 2, 3 and 4. Alternative splicing of this gene generates multiple transcript variants, but the full length nature of all the other variants but one has not been defined. [provided by RefSeq]

Sequence: MPGKLLWGDIMELEAPLEESESQKKERQKSDRRKSRHHYDSDEKSETRENGVTDDLDAPKAKKSKMKEKLNGDTEEGFNRLSDEFSKSHKSRRKDLPNGDIDEYEKKSKRVSSLDTSTHKSSDNKLEETLTREQKEGAFSNFPISEETIKLLKGRGVTYLFPIQVKTFGPVYEGKDLIAQARTGTGKTFSFAIPLIERLQRNQETIKKSRSPKVLVLAPTRELANQVAKDFKDITRKLSVACFYGGTSYQSQINHIRNGIDILVGTPGRIKDHLQSGRLDLSKLRHVVLDEVDQMLDLGFAEQVEDIIHESYKTDSEDNPQTLLFSATCPQWVYKVAKKYMKSRYEQVDLVGKMTQKAATTVEHLAIQCHWSQRPAVIGDVLQVYSGSEGRAIIFCETKKNVTEMAMNPHIKQNAQCLHGDIAQSQREITLKGFREGSFKVLVATNVAARGLDIPEVDLVIQSSPPQDVESYIHRSGRTGRAGRTGICICFYQPRERGQLRYVEQKAGITFKRVGVPSTMDLVKSKSMDAIRSLASVSYAAVDFFRPSAQRLIEEKGAVDALAAALAHISGASSFEPRSLITSDKGFVTMTLESLEEIQDVSCAWKELNRKLSSNAVSQITRMCLLKGNMGVCFDVPTTESERLQAEWHDSDWILSVPAKLPEIEEYYDGNTSSNSRQRSGWSSGRSGRSGRSGGRSGGRSGRQSRQGSRSGSRQDGRRRSGNRNRSRSGGHKRSFD
Specifications

Specifications

NP_076950.1
Liquid
79009
DDX50 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MPGKLLWGDIMELEAPLEESESQKKERQKSDRRKSRHHYDSDEKSETRENGVTDDLDAPKAKKSKMKEKLNGDTEEGFNRLSDEFSKSHKSRRKDLPNGDIDEYEKKSKRVSSLDTSTHKSSDNKLEETLTREQKEGAFSNFPISEETIKLLKGRGVTYLFPIQVKTFGPVYEGKDLIAQARTGTGKTFSFAIPLIERLQRNQETIKKSRSPKVLVLAPTRELANQVAKDFKDITRKLSVACFYGGTSYQSQINHIRNGIDILVGTPGRIKDHLQSGRLDLSKLRHVVLDEVDQMLDLGFAEQVEDIIHESYKTDSEDNPQTLLFSATCPQWVYKVAKKYMKSRYEQVDLVGKMTQKAATTVEHLAIQCHWSQRPAVIGDVLQVYSGSEGRAIIFCETKKNVTEMAMNPHIKQNAQCLHGDIAQSQREITLKGFREGSFKVLVATNVAARGLDIPEVDLVIQSSPPQDVESYIHRSGRTGRAGRTGICICFYQPRERGQLRYVEQKAGITFKRVGVPSTMDLVKSKSMDAIRSLASVSYAAVDFFRPSAQRLIEEKGAVDALAAALAHISGASSFEPRSLITSDKGFVTMTLESLEEIQDVSCAWKELNRKLSSNAVSQITRMCLLKGNMGVCFDVPTTESERLQAEWHDSDWILSVPAKLPEIEEYYDGNTSSNSRQRSGWSSGRSGRSGRSGGRSGGRSGRQSRQGSRSGSRQDGRRRSGNRNRSRSGGHKRSFD
RUO
DDX50
wheat germ expression system
Yes
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
109kDa
Glutathione Sepharose 4 Fast Flow
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
GU2|GUB|MGC3199|RH-II/GuB
DDX50
Wheat Germ (in vitro)
GST
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit