Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human DDX55 Full-length ORF (AAH35911.1, 1 a.a. - 207 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Manufacturer:  Abnova™ H00057696P01L

 View more versions of this product

Catalog No. 89-941-709


Add to cart

Description

Description

This gene encodes a member of the DEAD box protein family. DEAD box proteins, characterized by the conserved motif Asp-Glu-Ala-Asp (DEAD), are putative RNA helicases. They are implicated in a number of cellular processes involving alteration of RNA secondary structure, such as translation initiation, nuclear and mitochondrial splicing, and ribosome and spliceosome assembly. Based on their distribution patterns, some members of this family are believed to be involved in embryogenesis, spermatogenesis, and cellular growth and division. Multiple alternatively spliced transcript variants have been found for this gene, but the biological validity of only one transcript has been confirmed. [provided by RefSeq]

Sequence: MKPQRNTADLLPKLKSMALADRAVFEKGMKAFVSYVQAYAKHECNLIFRLKDLDFASLARGFALLRMPKMPELRGKQFPDFVPVDVNTDTIPFKDKIREKQRQKLLEQQRREKTENEGRRKFIKNKAWSKQKAKKEKKKKMNEKRKREEGSDIEDEDMEELLNDTRLLKKLKKGKITEEEFEKGLLTTGKRTIKTVDLGISDLEDDC
Specifications

Specifications

AAH35911.1
Liquid
57696
DDX55 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MKPQRNTADLLPKLKSMALADRAVFEKGMKAFVSYVQAYAKHECNLIFRLKDLDFASLARGFALLRMPKMPELRGKQFPDFVPVDVNTDTIPFKDKIREKQRQKLLEQQRREKTENEGRRKFIKNKAWSKQKAKKEKKKKMNEKRKREEGSDIEDEDMEELLNDTRLLKKLKKGKITEEEFEKGLLTTGKRTIKTVDLGISDLEDDC
RUO
DDX55
wheat germ expression system
Yes
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
50.7kDa
Glutathione Sepharose 4 Fast Flow
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
FLJ16577|KIAA1595|MGC33209
DDX55
Wheat Germ (in vitro)
GST
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit