Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human DEDD Full-length ORF (NP_001034800.1, 1 a.a. - 318 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Manufacturer:  Abnova™ H00009191P01

Catalog No. 89-941-743


Add to cart

Description

Description

This gene encodes a protein that contains a death effector domain (DED). DED is a protein-protein interaction domain shared by adaptors, regulators and executors of the programmed cell death pathway. Overexpression of this gene was shown to induce weak apoptosis. Upon stimulation, this protein was found to translocate from cytoplasm to nucleus and colocalize with UBTF, a basal factor required for RNA polymerase I transcription, in the nucleolus. At least three transcript variants encoding the same protein have been found for this gene. [provided by RefSeq]

Sequence: MAGLKRRASQVWPEEHGEQEHGLYSLHRMFDIVGTHLTHRDVRVLSFLFVDVIDDHERGLIRNGRDFLLALERQGRCDESNFRQVLQLLRIITRHDLLPYVTLKRRRAVCPDLVDKYLEETSIRYVTPRALSDPEPRPPQPSKTVPPHYPVVCCPTSGPQMCSKRPARGRATLGSQRKRRKSVTPDPKEKQTCDIRLRVRAEYCQHETALQGNVFSNKQDPLERQFERFNQANTILKSRDLGSIICDIKFSELTYLDAFWRDYINGSLLEALKGVFITDSLKQAVGHEAIKLLVNVDEEDYELGRQKLLRNLMLQALP
Specifications

Specifications

NP_001034800.1
Liquid
9191
DEDD (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MAGLKRRASQVWPEEHGEQEHGLYSLHRMFDIVGTHLTHRDVRVLSFLFVDVIDDHERGLIRNGRDFLLALERQGRCDESNFRQVLQLLRIITRHDLLPYVTLKRRRAVCPDLVDKYLEETSIRYVTPRALSDPEPRPPQPSKTVPPHYPVVCCPTSGPQMCSKRPARGRATLGSQRKRRKSVTPDPKEKQTCDIRLRVRAEYCQHETALQGNVFSNKQDPLERQFERFNQANTILKSRDLGSIICDIKFSELTYLDAFWRDYINGSLLEALKGVFITDSLKQAVGHEAIKLLVNVDEEDYELGRQKLLRNLMLQALP
RUO
DEDD
wheat germ expression system
Yes
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
63.2kDa
Glutathione Sepharose 4 Fast Flow
2 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
CASP8IP1|DEDD1|DEFT|FLDED1|KE05
DEDD
Wheat Germ (in vitro)
GST
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit