Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human DEDD Partial ORF (NP_127491, 91 a.a. - 190 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Manufacturer:  Abnova™ H00009191Q01S

 View more versions of this product

Catalog No. 89-941-741


Add to cart

Description

Description

This gene encodes a protein that contains a death effector domain (DED). DED is a protein-protein interaction domain shared by adaptors, regulators and executors of the programmed cell death pathway. Overexpression of this gene was shown to induce weak apoptosis. Upon stimulation, this protein was found to translocate from cytoplasm to nucleus and colocalize with UBTF, a basal factor required for RNA polymerase I transcription, in the nucleolus. At least three transcript variants encoding the same protein have been found for this gene. [provided by RefSeq]

Sequence: IITRHDLLPYVTLKRRRAVCPDLVDKYLEETSIRYVTPRALSDPEPRPPQPSKTVPPHYPVVCCPTSGPQMCSKRPARGRATLGSQRKRRKSVTPDPKEK
Specifications

Specifications

NP_127491
Liquid
9191
DEDD (Human) Recombinant Protein (Q01)
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
CASP8IP1|DEDD1|DEFT|FLDED1|KE05
DEDD
Wheat Germ (in vitro)
GST
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.74kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
IITRHDLLPYVTLKRRRAVCPDLVDKYLEETSIRYVTPRALSDPEPRPPQPSKTVPPHYPVVCCPTSGPQMCSKRPARGRATLGSQRKRRKSVTPDPKEK
RUO
DEDD
wheat germ expression system
Yes
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit