Learn More
Abnova™ Human DEFA1 (P59665) Partial Recombinant Protein
Used for Func, SDS-PAGE
Manufacturer:  Abnova™ P6122
Catalog No. 89-941-748
Add to cart
Description
Description
Defensins are a family of microbicidal and cytotoxic peptides thought to be involved in host defense. They are abundant in the granules of neutrophils and also found in the epithelia of mucosal surfaces such as those of the intestine, respiratory tract, urinary tract, and vagina. Members of the defensin family are highly similar in protein sequence and distinguished by a conserved cysteine motif. Several alpha defensin genes appear to be clustered on chromosome 8. The protein encoded by this gene, defensin, alpha 1, is found in the microbicidal granules of neutrophils and likely plays a role in phagocyte-mediated host defense. It differs from defensin, alpha 3 by only one amino acid. [provided by RefSeq]
Sequence: ACYCRIPACIAGERRYGYCIYQGRLWAFCC