Learn More
Abnova™ Human DEFB4 (O15263, 41 amino acids) Partial Recombinant Protein
Used for Func, SDS-PAGE
Manufacturer:  Abnova™ P4067
Catalog No. 89-941-772
Add to cart
Description
Description
Defensins form a family of microbicidal and cytotoxic peptides made by neutrophils. Members of the defensin family are highly similar in protein sequence. This gene encodes defensin, beta 4, an antibiotic peptide which is locally regulated by inflammation. [provided by RefSeq]