Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human DEFB4 Partial ORF (NP_004933, 24 a.a. - 64 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Manufacturer:  Abnova™ H00001673Q01S

 View more versions of this product

Catalog No. 89-941-770


Add to cart

Description

Description

Defensins form a family of microbicidal and cytotoxic peptides made by neutrophils. Members of the defensin family are highly similar in protein sequence. This gene encodes defensin, beta 4, an antibiotic peptide which is locally regulated by inflammation. [provided by RefSeq]

Sequence: GIGDPVTCLKSGAICHPVFCPRRYKQIGTCGLPGTKCCKKP
Specifications

Specifications

NP_004933
Liquid
1673
DEFB4 (Human) Recombinant Protein (Q01)
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
DEFB-2|DEFB102|DEFB2|HBD-2|SAP1
DEFB4
Wheat Germ (in vitro)
GST
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
30.25kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
GIGDPVTCLKSGAICHPVFCPRRYKQIGTCGLPGTKCCKKP
RUO
DEFB4
wheat germ expression system
Yes
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit