Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human DEGS1 Full-length ORF (NP_003667.1, 1 a.a. - 323 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Manufacturer:  Abnova™ H00008560P01S

 View more versions of this product

Catalog No. 89-941-778


Add to cart

Description

Description

This gene encodes a member of the membrane fatty acid desaturase family which is responsible for inserting double bonds into specific positions in fatty acids. This protein contains three His-containing consensus motifs that are characteristic of a group of membrane fatty acid desaturases. It is predicted to be a multiple membrane-spanning protein localized to the endoplasmic reticulum. Overexpression of this gene inhibited biosynthesis of the EGF receptor, suggesting a possible role of a fatty acid desaturase in regulating biosynthetic processing of the EGF receptor. Two splice variants have been identified. [provided by RefSeq]

Sequence: MGSRVSREDFEWVYTDQPHADRRREILAKYPEIKSLMKPDPNLIWIIIMMVLTQLGAFYIVKDLDWKWVIFGAYAFGSCINHSMTLAIHEIAHNAAFGNCKAMWNRWFGMFANLPIGIPYSISFKRYHMDHHRYLGADGVDVDIPTDFEGWFFCTAFRKFIWVILQPLFYAFRPLFINPKPITYLEVINTVAQVTFDILIYYFLGIKSLVYMLAASLLGLGLHPISGHFIAEHYMFLKGHETYSYYGPLNLLTFNVGYHNEHHDFPNIPGKSLPLVRKIAAEYYDNLPHYNSWIKVLYDFVMDDTISPYSRMKRHQKGEMVLE
Specifications

Specifications

NP_003667.1
Liquid
8560
DEGS1 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MGSRVSREDFEWVYTDQPHADRRREILAKYPEIKSLMKPDPNLIWIIIMMVLTQLGAFYIVKDLDWKWVIFGAYAFGSCINHSMTLAIHEIAHNAAFGNCKAMWNRWFGMFANLPIGIPYSISFKRYHMDHHRYLGADGVDVDIPTDFEGWFFCTAFRKFIWVILQPLFYAFRPLFINPKPITYLEVINTVAQVTFDILIYYFLGIKSLVYMLAASLLGLGLHPISGHFIAEHYMFLKGHETYSYYGPLNLLTFNVGYHNEHHDFPNIPGKSLPLVRKIAAEYYDNLPHYNSWIKVLYDFVMDDTISPYSRMKRHQKGEMVLE
RUO
DEGS1
wheat germ expression system
Yes
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
64.3kDa
Glutathione Sepharose 4 Fast Flow
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
DEGS|DES1|Des-1|FADS7|MGC5079|MIG15|MLD
DEGS1
Wheat Germ (in vitro)
GST
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit