Learn More
Abnova™ Human DENR Full-length ORF (NP_003668.2, 1 a.a. - 198 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Supplier: Abnova™ H00008562P01L
Description
This gene encodes a protein whose expression was found to increase in cultured cells at high density but not during growth arrest. This gene was also shown to have increased expression in cells overexpressing HER-2/neu proto-oncogene. The protein contains an SUI1 domain. In budding yeast, SUI1 is a translation initiation factor that along with eIF-2 and the initiator tRNA-Met, directs the ribosome to the proper translation start site. Proteins similar to SUI have been found in mammals, insects, and plants. [provided by RefSeq]
Sequence: MAADISESSGADCKGDPRNSAKLDADYPLRVLYCGVCSLPTEYCEYMPDVAKCRQWLEKNFPNEFAKLTVENSPKQEAGISEGQGTAGEEEEKKKQKRGGRGQIKQKKKTVPQKVTIAKIPRAKKKYVTRVCGLATFEIDLKEAQRFFAQKFSCGASVTGEDEIIIQGDFTDDIIDVIQEKWPEVDDDSIEDLGEVKKSpecifications
NP_003668.2 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
48.5kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
DRP/DRP1/SMAP-3 | |
DENR | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
8562 | |
DENR (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MAADISESSGADCKGDPRNSAKLDADYPLRVLYCGVCSLPTEYCEYMPDVAKCRQWLEKNFPNEFAKLTVENSPKQEAGISEGQGTAGEEEEKKKQKRGGRGQIKQKKKTVPQKVTIAKIPRAKKKYVTRVCGLATFEIDLKEAQRFFAQKFSCGASVTGEDEIIIQGDFTDDIIDVIQEKWPEVDDDSIEDLGEVKK | |
RUO | |
DENR | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.