Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human DERA (NP_057038, 1 a.a. - 318 a.a. ) Full-length Recombinant Protein with His tag

Used for SDS-PAGE

Manufacturer:  Abnova™ P3947

Catalog No. 89-941-814


Add to cart

Description

Description

Sequence: MGSSHHHHHHSSGLVPRGSHMSAHNRGTELDLSWISKIQVNHPAVLRRAEQIQARRTVKKEWQAAWLLKAVTFIDLTTLSGDDTSSNIQRLCYKAKYPIREDLLKALNMHDKGITTAAVCVYPARVCDAVKALKAAGCNIPVASVAAGFPAGQTHLKTRLEEIRLAVEDGATEIDVVINRSLVLTGQWEALYDEIRQFRKACGEAHLKTILATGELGTLTNVYKASMIAMMAGSDFIKTSTGKETVNATFPVAIVMLRAIRDFFWKTGNKIGFKPAGGIRSAKDSLAWLSLVKEELGDEWLKPELFRIGASTLLSDIERQIYHHVTGRYAAYHDLPMS
Specifications

Specifications

NP_057038
Sodium Dodecyl Sulfate–Polyacrylamide Gel Electrophoresis
Liquid
37.3kDa
Escherichia coli expression system
>90% by SDS-PAGE
50 ug
Store at 4°C for 1-2 weeks. For long term storage store at -20°C or -80°C. Aliquot to avoid repeated freezing and thawing.
CGI-26|DEOC
DERA
Escherichia coli
His
0.5 mg/mL
Liquid
51071
DERA (Human) Recombinant Protein
Conventional Chromatography
Loading 3 ug protein in 15% SDS PAGE.
MGSSHHHHHHSSGLVPRGSHMSAHNRGTELDLSWISKIQVNHPAVLRRAEQIQARRTVKKEWQAAWLLKAVTFIDLTTLSGDDTSSNIQRLCYKAKYPIREDLLKALNMHDKGITTAAVCVYPARVCDAVKALKAAGCNIPVASVAAGFPAGQTHLKTRLEEIRLAVEDGATEIDVVINRSLVLTGQWEALYDEIRQFRKACGEAHLKTILATGELGTLTNVYKASMIAMMAGSDFIKTSTGKETVNATFPVAIVMLRAIRDFFWKTGNKIGFKPAGGIRSAKDSLAWLSLVKEELGDEWLKPELFRIGASTLLSDIERQIYHHVTGRYAAYHDLPMS
RUO
DERA
Escherichia coli expression system
Yes
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit